
tert-Butylchlorodimethylsilane solution

Synonym(s):
tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMSCl solution
Linear Formula:
(CH3)3CSi(CH3)2Cl
CAS Number:
Molecular Weight:
150.72
Products
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing