Skip to Content
Merck
TentaGel™ S-NH2 extent of labeling: 0.20-0.35 mmol/g N loading

TentaGel S-NH2

TentaGel™ S-NH2 extent of labeling: 0.20-0.35 mmol/g N loading
Synonym(s):
O-(2-Aminoethyl)polyethylene glycol polymer-bound

Products

Brand
Physical Form
Product Line
Quality Segment
Functional Group
Application
Particle Size (µm)
Chain Type
Reaction Type