
sn-Glycerol 3-phosphate bis(cyclohexylammonium) salt

Synonym(s):
L-α-Glycerol phosphate bis(cyclohexylammonium) salt, L-α-Glycerophosphate bis(cyclohexylammonium) salt
Empirical Formula (Hill Notation):
C3H9O6P · 2C6H13N
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing