
Quetiapine Related Compound H

Synonym(s):
Quetiapine N-oxide, 2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-oxido-1-piperazinyl)ethoxy]ethanol, 4-(Dibenzo[b,f][1,4]thiazepin-11-yl)-1-[2-(2-hydroxyethoxy)ethyl]piperazine 1-oxide
Empirical Formula (Hill Notation):
C21H25N3O3S
CAS Number:
Molecular Weight:
399.51
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2