
Pymetrozin

Empirical Formula (Hill Notation):
C10H11N5O
CAS Number:
Molecular Weight:
217.23
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2