
Pramipexole Related Compound A

Synonym(s):
2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole
Empirical Formula (Hill Notation):
C7H11N3S
CAS Number:
Molecular Weight:
169.25
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2