Poly(allylamine hydrochloride)
Synonym(s):
PAA HCl, PAH, Poly(allylamine hydrochloride)
Linear Formula:
[CH2CH(CH2NH2 · HCl)]n
CAS Number:
Products
Brand
Manufacturer
Physical Form
Format
Quality Segment
Application
Polymer Composition
Polymer Type
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3