Skip to Content
Merck
Phloxine B antibacterial fluorescent dye

Phloxine B

Phloxine B antibacterial fluorescent dye
Synonym(s):
2′,4′,5′,7′-Tetrabromo-4,5,6,7-tetrachlorofluorescein disodium salt, Acid Red 92, Cyanosine, D&C;Red No. 28, Eosin 10B
Empirical Formula (Hill Notation):
C20H2Br4Cl4Na2O5
CAS Number:
Molecular Weight:
829.63
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing