
Phenothrin

Synonym(s):
Phenothrin
Empirical Formula (Hill Notation):
C23H26O3
CAS Number:
Molecular Weight:
350.45
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing