
N-Nitrosomethylethylamine

Synonym(s):
N-Nitroso-N-methylethylamine, NMEA
Empirical Formula (Hill Notation):
C3H8N2O
CAS Number:
Molecular Weight:
88.11
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2