
Nimesulide for peak identification

Synonym(s):
Nimesulide, N-(4-Nitro-2-phenoxyphenyl)methanesulfonamide
Empirical Formula (Hill Notation):
C13H12N2O5S
CAS Number:
Molecular Weight:
308.31
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing