Skip to Content
Merck
vibrant-m

Nevirapine Related Compound C

vibrant-m
Synonym(s):
5,11-Dihydro-4-methyl-11-propyl-6H-dipyrido[3,2-b:2′,3′-e][1,4]diazepin-6-one
Empirical Formula (Hill Notation):
C15H16N4O
CAS Number:
Molecular Weight:
268.31

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1