
Nα-Acetyl-L-lysine methyl ester hydrochloride

Linear Formula:
H2N(CH2)4CH(NHCOCH3)CO2CH3·HCl
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing