
Minocycline hydrochloride

Synonym(s):
Minocycline hydrochloride
Empirical Formula (Hill Notation):
C23H27N3O7 · HCl
CAS Number:
Molecular Weight:
493.94
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Research Area
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing