
Malondialdehyde tetrabutylammonium salt

Synonym(s):
Malonaldehyde tetrabutylammonium salt, Tetrabutylammonium malondialdehyde enolate
Linear Formula:
OCHCH=CHO[N(CH2CH2CH2CH3)4]
CAS Number:
Molecular Weight:
313.52
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing