Skip to Content
Merck
vibrant-m

Kromasil® Eternity C18 HPLC Column

vibrant-m

Products

Product Category
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (mm)
Application
Particle Size (µm)
Pore Size (Å)
Technique