
Isoxanthohumol

Synonym(s):
Isoxanthohumol, 2,3-Dihydro-7-hydroxy-2-(4-hydroxyphenyl)-5-methoxy-8-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one
Empirical Formula (Hill Notation):
C21H22O5
CAS Number:
Molecular Weight:
354.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing