
(−)-Isoproterenol hydrochloride

Synonym(s):
(−)-Isoprenaline hydrochloride, (−)-N-Isopropyl-L-noradrenaline hydrochloride, (R)-3,4-Dihydroxy-α-(isopropylaminomethyl)benzyl alcohol hydrochloride
Linear Formula:
3,4-(HO)2C6H3CH(OH)CH2NHCH(CH3)2·HCl
CAS Number:
Molecular Weight:
247.72
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Research Area
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing