
Hydromorphone HCl

Synonym(s):
Hydromorphone hydrochloride
Empirical Formula (Hill Notation):
C17H19NO3 · HCl
CAS Number:
Molecular Weight:
321.80
Products
Brand
Formula Weight
Physical Form
Agency
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing