
Helveticoside

Synonym(s):
Alleoside A, Deglucoerysimoside, Erisimin, Erysimotoxin, Strophanthidin 3β-digitoxoside
Empirical Formula (Hill Notation):
C29H42O9
CAS Number:
Molecular Weight:
534.64
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing