
Glycerol-2-13C

Synonym(s):
1,2,3-Propanetriol-2-13C, 13C Labeled glycerol, Glycerin-2-13C
Linear Formula:
(HOCH2)213CHOH
CAS Number:
Molecular Weight:
93.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing