
Formothion solution

Synonym(s):
S-[2-(Formylmethylamino)-2-oxoethyl] O,O-dimethyl dithiophosphate
Empirical Formula (Hill Notation):
C6H12NO4PS2
CAS Number:
Molecular Weight:
257.27
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing