
Fluorescein di(β-D-galactopyranoside)

Synonym(s):
FDG
Empirical Formula (Hill Notation):
C32H32O15
CAS Number:
Molecular Weight:
656.59
Products
Product Category
Brand
Formula Weight
Physical Form
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing