
Fenamiphos

Synonym(s):
Phenamiphos
Empirical Formula (Hill Notation):
C13H22NO3PS
CAS Number:
Molecular Weight:
303.36
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2