Skip to Content
Merck
Ethofumesate PESTANAL®, analytical standard

Ethofumesate

Ethofumesate PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C13H18O5S
CAS Number:
Molecular Weight:
286.34
EC Number:

Products

Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing