Skip to Content
Merck
Erythrosin extra bluish certified by the BSC

Erythrosin extra bluish

Erythrosin extra bluish certified by the BSC
Synonym(s):
2′,4′,5′,7′-Tetraiodofluorescein disodium salt, Acid Red 51, Iodoeosin
Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Agency
Chemical Composition
Application
Technique