
EPN

Synonym(s):
O-Ethyl O-(4-nitrophenyl) phenylphosphonothioate
Empirical Formula (Hill Notation):
C14H14NO4PS
CAS Number:
Molecular Weight:
323.30
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing