
DMT-2′O-TC-rC(ac) Phosphoramidite

Synonym(s):
N4-Acetyl-O5′-(4,4′-dimethoxytrityl)-O2′-(1,1-dioxothiomorpholine-4-thiocarbonyl)cytidine O3′-(O-(2-cyanoethyl)-N,N-diisopropylphosphoramidite)
Empirical Formula (Hill Notation):
C46H57N6O11PS2
CAS Number:
Molecular Weight:
965.08
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing