
Diisopropyl ether

Synonym(s):
Isopropyl ether, Diisopropyl ether
Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-12 of 12
Compare
Product Number
Description
Pricing