Skip to Content
Merck
Dehydroisoandrosterone 3-sulfate sodium salt dihydrate ≥93% (TLC), powder

Dehydroisoandrosterone 3-sulfate sodium salt dihydrate

Dehydroisoandrosterone 3-sulfate sodium salt dihydrate ≥93% (TLC), powder
Synonym(s):
5-Androsten-3β-ol-17-one sulfate sodium salt dihydrate, Prasterone sulfate sodium salt dihydrate, Sodium dehydroepiandrosterone sulfate
Empirical Formula (Hill Notation):
C19H27NaO5S · 2H2O
CAS Number:
Molecular Weight:
426.50
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing