
Cyproheptadine Related Compound C

Synonym(s):
5-(1-Methylpiperidin-4-yl)-5H-dibenzo[a,d]cyclohepten-5-ol
Empirical Formula (Hill Notation):
C21H23NO
CAS Number:
Molecular Weight:
305.41
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing