Skip to Content
Merck
Chlorodicyclohexylborane solution 1 M in hexanes

Chlorodicyclohexylborane solution

Chlorodicyclohexylborane solution 1 M in hexanes
Synonym(s):
Dicyclohexylboron chloride, Dicyclohexylboryl chloride, Dicyclohexylchloroborane
Linear Formula:
(C6H11)2BCl
CAS Number:
Molecular Weight:
212.57

Products

Boiling Point (°C)
Formula Weight
Physical Form
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing