
Chenodeoxycholic acid for system suitability

Synonym(s):
Chenodeoxycholic acid, 3α,7α-Dihydroxy-5β-cholanic acid, 5β-Cholanic acid-3α,7α-diol, Chenodiol
Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing