![Cabergoline European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/866/727/520055ed-67f5-441b-896d-d68a3ee2ed2b/640/520055ed-67f5-441b-896d-d68a3ee2ed2b.png)
Cabergoline
![Cabergoline European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/866/727/520055ed-67f5-441b-896d-d68a3ee2ed2b/640/520055ed-67f5-441b-896d-d68a3ee2ed2b.png)
Synonym(s):
1-Ethyl-3-(3′-dimethylaminopropyl)-3-(6′-allylergoline-8′β-carbonyl)urea, 1-[(6-Allylergoline-8β-yl)carbonyl]-1-[3-(dimethylamino)-propyl]-3-ethylurea, Cabaser, Dostinex, FCE-21336, Cabergoline
Empirical Formula (Hill Notation):
C26H37N5O2
CAS Number:
Molecular Weight:
451.60
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3