
Anti-MED12 antibody produced in rabbit

Synonym(s):
Anti-ARC240 antibody produced in rabbit, Anti-Activator-recruited cofactor 240 kDa component antibody produced in rabbit, Anti-CAG repeat protein 45 antibody produced in rabbit, Anti-Mediator of RNA polymerase II transcription subunit 12 antibody produced in rabbit, Anti-OPA-containing protein antibody produced in rabbit, Anti-Thyroid hormone receptor-associated protein complex 230 kDa component antibody produced in rabbit, Anti-Trap230 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing