
Amoxicillin Related Compound C

Synonym(s):
(4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic acid, Amoxicillin diketopiperazine
Empirical Formula (Hill Notation):
C16H19N3O5S
Molecular Weight:
365.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2