
Aluminium potassium sulfate dodecahydrate

Synonym(s):
Aluminium potassium sulfate dodecahydrate
Linear Formula:
KAl(SO4)2 · 12 H2O
CAS Number:
Molecular Weight:
474.39
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-2 of 2