
Alosetron hydrochloride

Synonym(s):
2,3,4,5-Tetrahydro-5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-1H-pyrido[4,3-b]indol-1-one hydrochloride, Alosetron hydrochloride
Empirical Formula (Hill Notation):
C17H18N4O·HCl
CAS Number:
Molecular Weight:
330.81
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing