
Acrylate-PEG-NHS

Synonym(s):
Acrylate-PEG-NHS
Linear Formula:
CH2CHCOO(CH2CH2O)nCH2COOC4H4O2N
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reactivity
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing