
Acetyleugenol

Synonym(s):
Eugenyl acetate, 4-Allyl-2-methoxyphenyl acetate, O-Acetyleugenol, Eugenol acetate
Empirical Formula (Hill Notation):
C12H14O3
CAS Number:
Molecular Weight:
206.24
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3