
4-Dodecylresorcinol

Linear Formula:
CH3(CH2)11C6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
278.43
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing