
3-Methyl ellagic acid

Synonym(s):
3-O-Methylellagic acid, 2,3,7-Trihydroxy-8-methoxy-[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione, 2,7,8-Trihydroxy-3-methoxychromeno[5,4,3-cde]chromene-5,10-dione, Ellagic acid 3-methyl ether
Empirical Formula (Hill Notation):
C15H8O8
CAS Number:
Molecular Weight:
316.22
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing