Skip to Content
Merck
2-Butanol JIS special grade, ≥99.0%

2-Butanol

2-Butanol JIS special grade, ≥99.0%
Synonym(s):
sec-Butyl alcohol, 2-Butanol
Linear Formula:
CH3CH2CH(OH)CH3
CAS Number:
Molecular Weight:
74.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8