Skip to Content
Merck
1-Butyl-3-methylimidazolium methanesulfonate ≥95%

1-Butyl-3-methylimidazolium methanesulfonate

1-Butyl-3-methylimidazolium methanesulfonate ≥95%
Synonym(s):
1-Butyl-3-methyl-1H-imidazolium methanesulfonate
Empirical Formula (Hill Notation):
C9H18N2O3S
CAS Number:
Molecular Weight:
234.32

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing