Skip to Content
Merck
1,11-Dibromoundecane ≥98%

1,11-Dibromoundecane

1,11-Dibromoundecane ≥98%
Synonym(s):
Undecamethylene dibromide
Linear Formula:
Br(CH2)11Br
CAS Number:
Molecular Weight:
314.10
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing