Skip to Content
Merck
All Photos(1)

Key Documents

80229

Supelco

Phenyl benzoate

analytical standard

Synonym(s):

Benzoic acid phenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2C6H5
CAS Number:
Molecular Weight:
198.22
Beilstein:
1566346
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

298-299 °C (lit.)

mp

68-70 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

O=C(Oc1ccccc1)c2ccccc2

InChI

1S/C13H10O2/c14-13(11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h1-10H

InChI key

FCJSHPDYVMKCHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service