Skip to Content
Merck
All Photos(1)

Key Documents

Z710180

Nichipet Eco pipette

volume 20-200 μL

Synonym(s):

Eco pipette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

description

With glass tip

feature

electronic: no
single-channel

packaging

pack of 1 ea

manufacturer/tradename

Nichiryo 00-NPC-200

volume measuring range

20-200 μL

volume

200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Providing the accuracy and precision of a micropipette with the compatibility of glass. Glass tips allow pipetting of substances not compatibile with plastic.
  • Glass tip enables totally bias free liquid handling for endocrine disruptors analysis
  • 4 sizes cover a wide volume range (20-10000 μL)
  • Nozzle with double O-rings enables easy tip fitting/ejection
  • Reliable sealing nozzle is PTFE and piston is ceramic

Application

Nichipet Eco pipette is suitable for sample transfer from different environmental and food samples.

Features and Benefits

  • Micropipette provides accuracy and precision
  • Glass tips provide reliability and stability
  • Glass tips enable bias-free liquid handling for endocrine disruptor studies
  • The double-O-ring design enables the attachment of the glass tip to the micropipette
  • Made of Polytetrafluoroethylene (PTFE) nozzle and a ceramic piston


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service