Skip to Content
Merck
All Photos(1)

Key Documents

SML3494

Sigma-Aldrich

β-Benzamide adenine dinucleotide sodium salt

≥95% (HPLC)

Synonym(s):

Adenosine 5′-(trihydrogen diphosphate) P′→5′-ester with 3-β-D-ribofuranosylbenzamide sodium salt, BAD, benzamide adenine dinucleotide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N6O14P2Na2
Molecular Weight:
706.40
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

storage temp.

-10 to -25°C

SMILES string

O[C@@H]1[C@H](O)[C@@H](COP(OP(OC[C@@H]2[C@@H](O)[C@@H](O)[C@H](N(C=N3)C4=C3C(N)=NC=N4)O2)(O[Na])=O)(O[Na])=O)O[C@H]1C5=CC(C(N)=O)=CC=C5

Biochem/physiol Actions

β-Benzamide adenine dinucleotide (BAD), an analogue of nicotinamide adenine dinucleotide (β-NAD+), is an active metabolite of benzamide riboside and a potent inhibitor of inosine 5′-monophosphate dehydrogenase (IMPDH). Also, β-Benzamide adenine dinucleotide is a non-hydrolysable substrate/inhibitor of NADases and PARP enzymes. A higher concentration (Ki = 90 μM) it inhibits human NAD kinase.
Analogue of nicotinamide adenine dinucleotide (β-NAD+); inhibitor of inosine 5′-monophosphate dehydrogenase (IMPDH)

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service