Skip to Content
Merck
All Photos(1)

Key Documents

SML2660

Sigma-Aldrich

ABT-100 Hydrochloride

≥98% (HPLC)

Synonym(s):

6-[(2S)-2-(4-Cyanophenyl)-2-hydroxy-2-(1-methyl-1H-imidazol-5-yl)ethoxy]-4′-(trifluoromethoxy)-1,1′-biphenyl-3-carbonitrile hydrochloride, 6-[(2S)-2-(4-Cyanophenyl)-2-hydroxy-2-(1-methyl-1H-imidazol-5-yl)ethoxy]-4′-(trifluoromethoxy)[1,1′-biphenyl]-3-carbonitrile hydrochloride, A-409100 Hydrochloride, ABT 100, [1,1′-Biphenyl]-3-carbonitrile, 6-[(2S)-2-(4-cyanophenyl)-2-hydroxy-2-(1-methyl-1H-imidazol-5-yl)ethoxy]-4′-(trifluoromethoxy)- hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H19F3N4O3 · HCl
CAS Number:
Molecular Weight:
540.92
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@](COC1=C(C2=CC=C(OC(F)(F)F)C=C2)C=C(C=C1)C#N)(C3=CN=CN3C)C(C=C4)=CC=C4C#N.Cl

Biochem/physiol Actions

ABT-100 is an orally available, highly potent and highly selective inhibitor of farnesyltransferase (FTase). ABT-100 exhibits broad spectrum antitumor activities in xenograft models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service