Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400172

Sigma-Aldrich

Anti-MX2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MXB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MX2(4600)

Related Categories

General description

The protein encoded by this gene has a nuclear and a cytoplasmic form and is a member of both the dynamin family and the family of large GTPases. The nuclear form is localized in a granular pattern in the heterochromatin region beneath the nuclear envelope. A nuclear localization signal (NLS) is present at the amino terminal end of the nuclear form but is lacking in the cytoplasmic form due to use of an alternate translation start codon. This protein is upregulated by interferon-alpha but does not contain the antiviral activity of a similar myxovirus resistance protein 1. (provided by RefSeq)

Immunogen

MX2 (AAH35293, 1 a.a. ~ 715 a.a) full-length human protein.

Sequence
MSKAHKPWPYRRRSQFSSRKYLKKEMNSFQQQPPPFGTVPPQMMFPPNWQGAEKDAAFLAKDFNFLTLNNQPPPGNRSQPRAMGPENNLYSQYEQKVRPCIDLIDSLRALGVEQDLALPAIAVIGDQSSGKSSVLEALSGVALPRGSGIVTRCPLVLKLKKQPCEAWAGRISYRNTELELQDPGQVEKEIHKAQNVMAGNGRGISHELISLEITSPEVPDLTIIDLPGITRVAVDNQPRDIGLQIKALIKKYIQRQQTINLVVVPCNVDIATTEALSMAHEVDPEGDRTIGILTKPDLMDRGTEKSVMNVVRNLTYPLKKGYMIVKCRGQQEITNRLSLAEATKKEITFFQTHPYFRVLLEEGSATVPRLAERLTTELIMHIQKSLPLLEGQIRESHQKATEELRRCGADIPSQEADKMFFLIEKIKMFNQDIEKLVEGEEVVRENETRLYNKIREDFKNWVGILATNTQKVKNIIHEEVEKYEKQYRGKELLGFVNYKTFEIIVHQYIQQLVEPALSMLQKAMEIIQQAFINVAKKHFGEFFNLNQTVQSTIEDIKVKHTAKAENMIQLQFRMEQMVFCQDQIYSVVLKKVREEIFNPLGTPSQNMKLNSHFPSNESSVSSFTEIGIHLNAYFLETSKRLANQIPFIIQYFMLRENGDSLQKAMMQILQEKNRYSWLLQEQSETATKRRILKERIYRLTQARHALCQFSSKEIH

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claire Deleage et al.
Science advances, 5(5), eaav7116-eaav7116 (2019-06-01)
Understanding HIV transmission is critical to guide the development of prophylactic interventions to prevent infection. We used a nonhuman primate (NHP) model with a synthetic swarm of sequence-tagged variants of SIVmac239 ("SIVmac239X") and scheduled necropsy during primary infection (days 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service